Learn More
2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, 99%
CAS: 16357-59-8 | C14H17NO3 | 247.29 g/mol
$56.66 - $637.27
Chemical Identifiers
| CAS | 16357-59-8 |
|---|---|
| Molecular Formula | C14H17NO3 |
| Molecular Weight (g/mol) | 247.29 |
| MDL Number | MFCD00006703 |
| InChI Key | GKQLYSROISKDLL-UHFFFAOYNA-N |
| Synonym | eedq, 2-ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, n-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, ethyl 2-ethoxyquinoline-1 2h-carboxylate, 1-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 2-ethoxy-1 2h-quinolinecarboxylic acid, ethyl ester, 1 2h-quinolinecarboxylic acid, 2-ethoxy-, ethyl ester, ethyl 2-ethoxy-1,2-dihydroquinoline-1-carboxylate, ethyl 1,2-dihydro-2-ethoxy-1-quinolinecarboxylate, ethyl n-2-ethoxy-1,2-dihydroquinoline carboxylate |
| PubChem CID | 27833 |
| IUPAC Name | ethyl 2-ethoxy-2H-quinoline-1-carboxylate |
| SMILES | CCOC1C=CC2=CC=CC=C2N1C(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1372409
|
Thermo Scientific Chemicals
A1372409 |
10 g |
Each for $56.66
|
|
|||||
|
AAA1372418
|
Thermo Scientific Chemicals
A1372418 |
50 g |
Each for $180.05
|
|
|||||
|
AAA1372430
|
Thermo Scientific Chemicals
A1372430 |
250 g |
Each for $637.27
|
|
|||||
Description
2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline is an irreversible membrane-bound receptor antagonist. It is used in synthetic chemistry, discovery, process R&D. It also acts as coupling reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline is an irreversible membrane-bound receptor antagonist. It is used in synthetic chemistry, discovery, process R&D. It also acts as coupling reagents.
Solubility
Soluble in 95% Ethanol (50 mg/ml). Insoluble in water.
Notes
Moisture Sensitive. Store at 4°C. Incompatible with acids and strong oxidizing agents.
Chemical Identifiers
| 16357-59-8 | |
| 247.29 | |
| GKQLYSROISKDLL-UHFFFAOYNA-N | |
| 27833 | |
| CCOC1C=CC2=CC=CC=C2N1C(=O)OCC |
| C14H17NO3 | |
| MFCD00006703 | |
| eedq, 2-ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, n-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, ethyl 2-ethoxyquinoline-1 2h-carboxylate, 1-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 2-ethoxy-1 2h-quinolinecarboxylic acid, ethyl ester, 1 2h-quinolinecarboxylic acid, 2-ethoxy-, ethyl ester, ethyl 2-ethoxy-1,2-dihydroquinoline-1-carboxylate, ethyl 1,2-dihydro-2-ethoxy-1-quinolinecarboxylate, ethyl n-2-ethoxy-1,2-dihydroquinoline carboxylate | |
| ethyl 2-ethoxy-2H-quinoline-1-carboxylate |
Specifications
| 16357-59-8 | |
| 125°C to 128°C (0.1 mmHg) | |
| MFCD00006703 | |
| 533048 | |
| 14,3518 | |
| Soluble in 95% Ethanol (50mg/ml). Insoluble in water. | |
| CCOC1C=CC2=CC=CC=C2N1C(=O)OCC | |
| 247.29 | |
| 247.29 | |
| 2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline |
| 63°C to 66°C | |
| C14H17NO3 | |
| 10 g | |
| Moisture sensitive | |
| eedq, 2-ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, n-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, ethyl 2-ethoxyquinoline-1 2h-carboxylate, 1-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 2-ethoxy-1 2h-quinolinecarboxylic acid, ethyl ester, 1 2h-quinolinecarboxylic acid, 2-ethoxy-, ethyl ester, ethyl 2-ethoxy-1,2-dihydroquinoline-1-carboxylate, ethyl 1,2-dihydro-2-ethoxy-1-quinolinecarboxylate, ethyl n-2-ethoxy-1,2-dihydroquinoline carboxylate | |
| GKQLYSROISKDLL-UHFFFAOYNA-N | |
| ethyl 2-ethoxy-2H-quinoline-1-carboxylate | |
| 27833 | |
| 99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 240-418-2
RTECSNumber : VB2010000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only