missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, 99%

Catalog No. AAA1372409
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1372409 10 g
AAA1372418 50 g
AAA1372430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1372409 Supplier Thermo Scientific Chemicals Supplier No. A1372409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16357-59-8 | C14H17NO3 | 247.29 g/mol

2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline is an irreversible membrane-bound receptor antagonist. It is used in synthetic chemistry, discovery, process R&D. It also acts as coupling reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline is an irreversible membrane-bound receptor antagonist. It is used in synthetic chemistry, discovery, process R&D. It also acts as coupling reagents.

Solubility
Soluble in 95% Ethanol (50 mg/ml). Insoluble in water.

Notes
Moisture Sensitive. Store at 4°C. Incompatible with acids and strong oxidizing agents.

Chemical Identifiers

CAS 16357-59-8
Molecular Formula C14H17NO3
Molecular Weight (g/mol) 247.29
MDL Number MFCD00006703
InChI Key GKQLYSROISKDLL-UHFFFAOYNA-N
Synonym eedq, 2-ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, n-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, ethyl 2-ethoxyquinoline-1 2h-carboxylate, 1-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 2-ethoxy-1 2h-quinolinecarboxylic acid, ethyl ester, 1 2h-quinolinecarboxylic acid, 2-ethoxy-, ethyl ester, ethyl 2-ethoxy-1,2-dihydroquinoline-1-carboxylate, ethyl 1,2-dihydro-2-ethoxy-1-quinolinecarboxylate, ethyl n-2-ethoxy-1,2-dihydroquinoline carboxylate
PubChem CID 27833
IUPAC Name ethyl 2-ethoxy-2H-quinoline-1-carboxylate
SMILES CCOC1C=CC2=CC=CC=C2N1C(=O)OCC

Specifications

Melting Point 63°C to 66°C
Boiling Point 125°C to 128°C (0.1 mmHg)
Quantity 10 g
Beilstein 533048
Sensitivity Moisture sensitive
Merck Index 14,3518
Solubility Information Soluble in 95% Ethanol (50mg/ml). Insoluble in water.
Formula Weight 247.29
Percent Purity 99%
Chemical Name or Material 2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 240-418-2
RTECSNumber VB2010000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only