missing translation for 'onlineSavingsMsg'
Learn More

2-Dodecylthiophene 97.0+%, TCI America™

Catalog No. D48745ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48745ML 5 mL
D48741ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48745ML Supplier TCI America Supplier No. D48745ML
Only null left

Chemical Identifiers

CAS 4861-61-4
Molecular Formula C16H28S
Molecular Weight (g/mol) 252.46
MDL Number MFCD22493346
InChI Key NJPMFDNZCLKTHE-UHFFFAOYSA-N
PubChem CID 566347
IUPAC Name 2-dodecylthiophene
SMILES CCCCCCCCCCCCC1=CC=CS1

Specifications

Color Yellow
Boiling Point 155°C
Quantity 5 mL
Formula Weight 252.46
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Dodecylthiophene