accessibility menu, dialog, popup

UserName

H3CS

CAS RN 4861-61-4

IUPAC Name: 2-dodecylthiophene
Synonyms: 2-dodecylthiophene
Molecular Weight (g/mol): 252.46
Molecular Formula: C16H28S
InChi Key: NJPMFDNZCLKTHE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=CC=CS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results