missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Dodecanol 98.0+%, TCI America™
Supplier: TCI America D29605G
| Quantity | 5 g |
|---|
Identifiants chimiques
| 10203-28-8 | |
| 186.34 | |
| XSWSEQPWKOWORN-UHFFFAOYNA-N | |
| 25045 | |
| CCCCCCCCCCC(C)O |
| C12H26O | |
| MFCD00004551 | |
| 2-dodecanol, dodecanol-2, 2-dodecanol, r, 2-dodecanol, s, dodecanol 2, secondary dodecanol, acmc-20luta, acmc-1bxbj, acmc-20m2p3 | |
| dodecan-2-ol |
Spécifications
| 2-Dodecanol | |
| 19°C | |
| 129°C | |
| 5 g | |
| 2-dodecanol, dodecanol-2, 2-dodecanol, r, 2-dodecanol, s, dodecanol 2, secondary dodecanol, acmc-20luta, acmc-1bxbj, acmc-20m2p3 | |
| CCCCCCCCCCC(C)O | |
| 186.34 | |
| 186.34 | |
| Liquid |
| 10203-28-8 | |
| Yellow | |
| C12H26O | |
| MFCD00004551 | |
| XSWSEQPWKOWORN-UHFFFAOYNA-N | |
| dodecan-2-ol | |
| 25045 | |
| ≥98.0% (GC) |
Sécurité et manipulation
EINECSNumber : (2)-0217&(2)-3704
TSCA : No