accessibility menu, dialog, popup

UserName

Explore New Spotlight Deals on Equipment, Instruments, and More

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 10203-28-8

H3CCH3OH

CAS RN 10203-28-8

IUPAC Name: dodecan-2-ol
Synonyms: 2-dodecanol
Molecular Weight (g/mol): 186.34
Molecular Formula: C12H26O
InChi Key: XSWSEQPWKOWORN-UHFFFAOYNA-N
SMILES: CCCCCCCCCCC(C)O
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.