missing translation for 'onlineSavingsMsg'
Learn More

2-(Diphenylphosphino)benzaldehyde oxime, 95%, Thermo Scientific Chemicals

Catalog No. p-7034236
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5190306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5190306 Supplier Thermo Scientific Chemicals Supplier No. H5190306
Only null left

It is used in the synthesis, structure and applications to the catalytic rearrangement and dehydration of aldoximes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis, structure and applications to the catalytic rearrangement and dehydration of aldoximes.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 153358-05-5
Molecular Formula C19H16NOP
Molecular Weight (g/mol) 305.32
MDL Number MFCD16251585
InChI Key XHIVESUSSLEMGJ-UHFFFAOYSA-N
Synonym 2-diphenylphosphino benzaldehyde oxime, n-2-diphenylphosphino benzylidene hydroxylamine, e-n-2-diphenylphosphanyl phenyl methylidene hydroxylamine
PubChem CID 98043674
IUPAC Name (NZ)-N-[(2-diphenylphosphanylphenyl)methylidene]hydroxylamine
SMILES ON=CC1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 305.32
Percent Purity 95%
Chemical Name or Material 2-(Diphenylphosphino)benzaldehyde oxime
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only