missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Dimethylphenylsilyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95.0+%, TCI America™
Supplier: TCI America D43575G
Specifications
| 2-(Dimethylphenylsilyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| Yellow | |
| C14H23BO2Si | |
| 5 g | |
| ARMSAQNLTKGMGM-UHFFFAOYSA-N | |
| dimethyl-phenyl-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)silane | |
| 10199126 | |
| ≥95.0% (GC) |
| 185990-03-8 | |
| 120°C | |
| MFCD05664111 | |
| (Dimethylphenylsilyl)boronic Acid Pinacol Ester | |
| B1(OC(C(O1)(C)C)(C)C)[Si](C)(C)C2=CC=CC=C2 | |
| 262.231 | |
| 262.23 | |
| Liquid |
Chemical Identifiers
| 185990-03-8 | |
| 262.231 | |
| ARMSAQNLTKGMGM-UHFFFAOYSA-N | |
| 10199126 | |
| B1(OC(C(O1)(C)C)(C)C)[Si](C)(C)C2=CC=CC=C2 |
| C14H23BO2Si | |
| MFCD05664111 | |
| (Dimethylphenylsilyl)boronic Acid Pinacol Ester | |
| dimethyl-phenyl-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)silane |
Safety and Handling
TSCA : No