missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Dimethylphenylsilyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95.0+%, TCI America™
$201.32 - $665.46
Chemical Identifiers
| CAS | 185990-03-8 |
|---|---|
| Molecular Formula | C14H23BO2Si |
| Molecular Weight (g/mol) | 262.231 |
| MDL Number | MFCD05664111 |
| InChI Key | ARMSAQNLTKGMGM-UHFFFAOYSA-N |
| Synonym | (Dimethylphenylsilyl)boronic Acid Pinacol Ester |
| PubChem CID | 10199126 |
| IUPAC Name | dimethyl-phenyl-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)silane |
| SMILES | B1(OC(C(O1)(C)C)(C)C)[Si](C)(C)C2=CC=CC=C2 |
Chemical Identifiers
| 185990-03-8 | |
| 262.231 | |
| ARMSAQNLTKGMGM-UHFFFAOYSA-N | |
| 10199126 | |
| B1(OC(C(O1)(C)C)(C)C)[Si](C)(C)C2=CC=CC=C2 |
| C14H23BO2Si | |
| MFCD05664111 | |
| (Dimethylphenylsilyl)boronic Acid Pinacol Ester | |
| dimethyl-phenyl-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)silane |
Specifications
| 185990-03-8 | |
| 120°C | |
| MFCD05664111 | |
| (Dimethylphenylsilyl)boronic Acid Pinacol Ester | |
| B1(OC(C(O1)(C)C)(C)C)[Si](C)(C)C2=CC=CC=C2 | |
| 262.231 | |
| 262.23 | |
| Liquid |
| Yellow | |
| C14H23BO2Si | |
| 1 g | |
| ARMSAQNLTKGMGM-UHFFFAOYSA-N | |
| dimethyl-phenyl-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)silane | |
| 10199126 | |
| ≥95.0% (GC) | |
| 2-(Dimethylphenylsilyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No