missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylcarbamoyl)pyridine-5-boronic acid pinacol ester, 96%

Catalog No. p-7036337
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54614MD 250 mg
AAH5461403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH54614MD Supplier Thermo Scientific Chemicals Supplier No. H54614MD
Only null left

CAS: 1006876-27-2 | C14H21BN2O3 | 276.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1006876-27-2
Molecular Formula C14H21BN2O3
Molecular Weight (g/mol) 276.143
MDL Number MFCD08458484
InChI Key RNCGUUHRBRONRG-UHFFFAOYSA-N
Synonym n,n-dimethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl picolinamide, 6-dimethylcarbamoyl pyridin-3-ylboronic acid pinacol ester, n,n-dimethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carboxamide, 6-dimethylcarbamoyl pyridine-3-boronic acid pinacol ester, 2-dimethylcarbamoyl pyridine-5-boronic acid pinacol ester, n,n-dimethyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carboxamide, 2-n,n-dimthylaminocarbonyl pyridine-5-boronic acid pinacol ester, n,n-dimethyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl picolinamide, 2-pyridinecarboxamide, n,n-dimethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, n,n-dimethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinecarboxamide
PubChem CID 57416491
IUPAC Name N,N-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)C(=O)N(C)C

Specifications

Quantity 250 mg
Formula Weight 276.14
Percent Purity 96%
Chemical Name or Material 2-(Dimethylcarbamoyl)pyridine-5-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only