accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 1006876-27-2

BOOH3CCH3H3CH3CNONCH3H3C

CAS RN 1006876-27-2

IUPAC Name: N,N-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
Synonyms: N,N-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
Molecular Weight (g/mol): 276.14
Molecular Formula: C14H21BN2O3
InChi Key: RNCGUUHRBRONRG-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.