missing translation for 'onlineSavingsMsg'
Learn More

2-Dimethylaminomethylphenol (contains Phenol) 70.0+%, TCI America™

Catalog No. D065525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D065525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D065525ML Supplier TCI America Supplier No. D065525ML
Only null left

Chemical Identifiers

CAS 120-65-0
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00045793
InChI Key FUIQBJHUESBZNU-UHFFFAOYSA-N
PubChem CID 32897
IUPAC Name 2-[(dimethylamino)methyl]phenol
SMILES CN(C)CC1=CC=CC=C1O

Specifications

Quantity 25 mL
UN Number 1760
Formula Weight 151.21
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Dimethylaminomethylphenol (contains Phenol)
EINECSNumber (3)-0710
TSCA Yes