Learn More
2-Dimethylaminoisopropyl chloride hydrochloride, 98%, Thermo Scientific Chemicals
$106.36 - $106.36
Chemical Identifiers
| CAS | 4584-49-0 |
|---|---|
| Molecular Formula | C5H13Cl2N |
| Molecular Weight (g/mol) | 158.066 |
| MDL Number | MFCD00012534 |
| InChI Key | OCWGRWAYARCRTQ-UHFFFAOYSA-N |
| Synonym | 2-dimethylaminoisopropyl chloride hydrochloride, 2-chloro-1-dimethylamino propane hydrochloride, 2-chloro-n,n-dimethylpropan-1-amine hydrochloride, n,n-dimethyl-2-chloropropylamine hydrochloride, 2-chloropropyldimethylammonium chloride, 2-dimethylamino isopropyl chloride hydrochloride, 2-chloropropyldimethylamine hydrochloride, 2-chloropropyl-dimethylamine hydrochloride, 1-methyl-2-dimethylaminoethyl chloride hydrochloride, 2-chloropropyl dimethylamine hydrochloride |
| PubChem CID | 94294 |
| IUPAC Name | 2-chloro-N,N-dimethylpropan-1-amine;hydrochloride |
| SMILES | CC(CN(C)C)Cl.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2088236
|
Thermo Scientific Chemicals
B2088236 |
500 g |
Each for $106.36
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4584-49-0 | |
| 158.066 | |
| OCWGRWAYARCRTQ-UHFFFAOYSA-N | |
| 94294 | |
| CC(CN(C)C)Cl.Cl |
| C5H13Cl2N | |
| MFCD00012534 | |
| 2-dimethylaminoisopropyl chloride hydrochloride, 2-chloro-1-dimethylamino propane hydrochloride, 2-chloro-n,n-dimethylpropan-1-amine hydrochloride, n,n-dimethyl-2-chloropropylamine hydrochloride, 2-chloropropyldimethylammonium chloride, 2-dimethylamino isopropyl chloride hydrochloride, 2-chloropropyldimethylamine hydrochloride, 2-chloropropyl-dimethylamine hydrochloride, 1-methyl-2-dimethylaminoethyl chloride hydrochloride, 2-chloropropyl dimethylamine hydrochloride | |
| 2-chloro-N,N-dimethylpropan-1-amine;hydrochloride |
Specifications
| 4584-49-0 | |
| C5H13Cl2N | |
| 500 g | |
| Hygroscopic | |
| OCWGRWAYARCRTQ-UHFFFAOYSA-N | |
| 2-chloro-N,N-dimethylpropan-1-amine;hydrochloride | |
| 94294 | |
| 98% |
| 190°C to 193°C | |
| MFCD00012534 | |
| 3670215 | |
| 2-dimethylaminoisopropyl chloride hydrochloride, 2-chloro-1-dimethylamino propane hydrochloride, 2-chloro-n,n-dimethylpropan-1-amine hydrochloride, n,n-dimethyl-2-chloropropylamine hydrochloride, 2-chloropropyldimethylammonium chloride, 2-dimethylamino isopropyl chloride hydrochloride, 2-chloropropyldimethylamine hydrochloride, 2-chloropropyl-dimethylamine hydrochloride, 1-methyl-2-dimethylaminoethyl chloride hydrochloride, 2-chloropropyl dimethylamine hydrochloride | |
| CC(CN(C)C)Cl.Cl | |
| 158.066 | |
| 158.07 | |
| 2-Dimethylaminoisopropyl chloride hydrochloride |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 224-971-7
RTECSNumber : TY1225000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only