missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)ethyl Methacrylate (stabilized with MEHQ) 98.5+%, TCI America™

Catalog No. p-7131976
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M008225ML 25 mL
M0082100ML 100 mL
M0082500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M008225ML Supplier TCI America Supplier No. M008225ML
Only null left

Chemical Identifiers

CAS 2867-47-2
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00008589
InChI Key JKNCOURZONDCGV-UHFFFAOYSA-N
Synonym 2-dimethylamino ethyl methacrylate, dimethylaminoethyl methacrylate, ageflex fm-1, dmaema, usaf rh-3, 2-propenoic acid, 2-methyl-, 2-dimethylamino ethyl ester, 2-n,n-dimethylamino ethyl methacrylate, n,n-dimethylaminoethyl methacrylate, 2-dimethylaminoethyl methacrylate, n,n-dimethylethanolamine methacrylate
PubChem CID 17869
IUPAC Name 2-(dimethylamino)ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCN(C)C

Specifications

Melting Point -30°C
Color Colorless
Boiling Point 70°C
Quantity 25 mL
UN Number 2522
Formula Weight 157.21
Percent Purity ≥98.5% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Dimethylamino)ethyl Methacrylate (stabilized with MEHQ)
EINECSNumber (2)-1047&(2)-1048
RTECSNumber OZ4200000
TSCA Yes