accessibility menu, dialog, popup

UserName

H3CNCH3OOCH3CH2

CAS RN 2867-47-2

IUPAC Name: 2-(dimethylamino)ethyl 2-methylprop-2-enoate
Synonyms: dimethylaminoethyl methacrylate
Molecular Weight (g/mol): 157.21
Molecular Formula: C8H15NO2
InChi Key: JKNCOURZONDCGV-UHFFFAOYSA-N
SMILES: CN(C)CCOC(=O)C(C)=C
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)