missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)ethyl Methacrylate (stabilized with MEHQ) 98.5+%, TCI America™

Catalog No. m0082100ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
M0082100ML 100 mL
M008225ML 25 mL
M0082500ML 500 mL
3 options

Catalog No. M0082100ML

Supplier: TCI America M0082100ML

Chemical Identifiers

CAS 2867-47-2
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00008589
InChI Key JKNCOURZONDCGV-UHFFFAOYSA-N
Synonym 2-dimethylamino ethyl methacrylate, dimethylaminoethyl methacrylate, ageflex fm-1, dmaema, usaf rh-3, 2-propenoic acid, 2-methyl-, 2-dimethylamino ethyl ester, 2-n,n-dimethylamino ethyl methacrylate, n,n-dimethylaminoethyl methacrylate, 2-dimethylaminoethyl methacrylate, n,n-dimethylethanolamine methacrylate
PubChem CID 17869
IUPAC Name 2-(dimethylamino)ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCN(C)C

Specifications

Melting Point -30°C
Color Colorless
Boiling Point 70°C
Quantity 100 mL
UN Number 2522
Formula Weight 157.21
Percent Purity ≥98.5% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Dimethylamino)ethyl Methacrylate (stabilized with MEHQ)
EINECSNumber (2)-1047&(2)-1048
RTECSNumber OZ4200000
TSCA Yes