missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Dimethylamino)ethyl methacrylate, 97%, stab. with ca 0.2% 4-methoxyphenol
Description
2-(Dimethylamino)ethyl methacrylate is used in the synthesis of poly ((2-(Dimethylamino)ethyl methacrylate), block copolymers of 2-(Dimethylamino)ethyl methacrylate and n-butyl methacrylate and amphiphilic AB block copolymers of 2-(Dimethylamino)ethyl methacrylate. It is also used in the preparation of polymer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Dimethylamino)ethyl methacrylate is used in the synthesis of poly ((2-(Dimethylamino)ethyl methacrylate), block copolymers of 2-(Dimethylamino)ethyl methacrylate and n-butyl methacrylate and amphiphilic AB block copolymers of 2-(Dimethylamino)ethyl methacrylate. It is also used in the preparation of polymer.
Solubility
Soluble in water. (1.000 g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to moisture and light. Incompatible with oxidizing agents.
2-(Dimethylamino)ethyl methacrylate is used in the synthesis of poly ((2-(Dimethylamino)ethyl methacrylate), block copolymers of 2-(Dimethylamino)ethyl methacrylate and n-butyl methacrylate and amphiphilic AB block copolymers of 2-(Dimethylamino)ethyl methacrylate. It is also used in the preparation of polymer.
Solubility
Soluble in water. (1.000 g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to moisture and light. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | ∼-30°C |
| Density | 0.932 |
| Boiling Point | 186°C to 188°C |
| Flash Point | 70°C (158°F) |
| Refractive Index | 1.44 |
| Quantity | 100 mL |
| UN Number | UN2522 |
| Beilstein | 1757048 |
| Solubility Information | Soluble in water. (1.000g/L at 20°C). |
| Formula Weight | 157.21 |
| Show More |
Safety and Handling
| Hazard Category | H227-H302-H315-H317-H319-H330 |
| Hazard Statement | GHS H Statement H302-H312-H315-H319-H317-H227 Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. Combustible liquid. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P374-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 2-DIMETHYLAMINOETHYL METHACRYLATE |
| EINECSNumber | 220-688-8 |
| RTECSNumber | OZ4200000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only