Learn More
2-(Dimethylamino)ethyl acrylate, 98%, stab. with ca 0.1% 4-methoxyphenol
CAS: 2439-35-2 | C7H13NO2 | 143.19 g/mol
Supplier: Thermo Scientific Chemicals L13506AE
Description
2-(Dimethylamino)ethyl acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, as a catalytic agent, as a petrochemical additive and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(Dimethylamino)ethyl acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, as a catalytic agent, as a petrochemical additive and agrochemicals.
Solubility
Soluble in water (1.000 g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Sensitive to heat, light and moisture.
Specifications
| 2-(Dimethylamino)ethyl acrylate | |
| 2439-35-2 | |
| 0.943 | |
| 62°C (144°F) | |
| 1.438 | |
| MFCD00038233 | |
| UN3302 | |
| Light sensitive | |
| Soluble in water (1.000g/L at 20°C). | |
| CN(C)CCOC(=O)C=C | |
| 143.19 | |
| 143.19 |
| stabilized with ≈0.1% 4-methoxyphenol | |
| -60°C | |
| 64°C (12 mmHg) | |
| C7H13NO2 | |
| CH2=CHCO2CH2CH2N(CH3)2 | |
| 100 mL | |
| 1099119 | |
| 2-dimethylamino ethyl acrylate, dimethylaminoethyl acrylate, n,n-dimethylaminoethyl acrylate, adame, 2-propenoic acid, 2-dimethylamino ethyl ester, acrylic acid, 2-dimethylamino ethyl ester, 2-dimethylamino, unii-5yes121ftl, 2-n,n-dimethylamino ethyl acrylate, ageflex fa-1 | |
| DPBJAVGHACCNRL-UHFFFAOYSA-N | |
| 2-(dimethylamino)ethyl prop-2-enoate | |
| 17111 | |
| 98% |
Chemical Identifiers
| 2439-35-2 | |
| 143.19 | |
| DPBJAVGHACCNRL-UHFFFAOYSA-N | |
| 17111 | |
| CN(C)CCOC(=O)C=C |
| C7H13NO2 | |
| MFCD00038233 | |
| 2-dimethylamino ethyl acrylate, dimethylaminoethyl acrylate, n,n-dimethylaminoethyl acrylate, adame, 2-propenoic acid, 2-dimethylamino ethyl ester, acrylic acid, 2-dimethylamino ethyl ester, 2-dimethylamino, unii-5yes121ftl, 2-n,n-dimethylamino ethyl acrylate, ageflex fa-1 | |
| 2-(dimethylamino)ethyl prop-2-enoate |
Safety and Handling
GHS H Statement
H311-H330-H314-H318-H227-H302-H317
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
May cause an allergic skin reaction.
P210-P235-P260-P262-P264b-P270-P271-P272-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
H227-H302-H310+H330-H314-H317-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 2-DIMETHYLAMINOETHYL ACRYLATE
EINECSNumber : 219-460-0
RTECSNumber : AS8578000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only