missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)ethyl acrylate, 98%, stab. with ca 0.1% 4-methoxyphenol

Catalog No. p-7046223
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13506AC 25 mL
AAL13506AE 100 mL
AAL13506AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL13506AC Supplier Thermo Scientific Chemicals Supplier No. L13506AC

CAS: 2439-35-2 | C7H13NO2 | 143.19 g/mol

2-(Dimethylamino)ethyl acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, as a catalytic agent, as a petrochemical additive and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Dimethylamino)ethyl acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, as a catalytic agent, as a petrochemical additive and agrochemicals.

Solubility
Soluble in water (1.000 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Sensitive to heat, light and moisture.

Chemical Identifiers

CAS 2439-35-2
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00038233
InChI Key DPBJAVGHACCNRL-UHFFFAOYSA-N
Synonym 2-dimethylamino ethyl acrylate, dimethylaminoethyl acrylate, n,n-dimethylaminoethyl acrylate, adame, 2-propenoic acid, 2-dimethylamino ethyl ester, acrylic acid, 2-dimethylamino ethyl ester, 2-dimethylamino, unii-5yes121ftl, 2-n,n-dimethylamino ethyl acrylate, ageflex fa-1
PubChem CID 17111
IUPAC Name 2-(dimethylamino)ethyl prop-2-enoate
SMILES CN(C)CCOC(=O)C=C

Specifications

Melting Point -60°C
Density 0.943
Boiling Point 64°C (12 mmHg)
Flash Point 62°C (144°F)
Refractive Index 1.438
Linear Formula CH2=CHCO2CH2CH2N(CH3)2
Quantity 25 mL
UN Number UN3302
Beilstein 1099119
Sensitivity Light sensitive
Solubility Information Soluble in water (1.000g/L at 20°C).
Formula Weight 143.19
Percent Purity 98%
Chemical Name or Material 2-(Dimethylamino)ethyl acrylate, stabilized with ≈0.1% 4-methoxyphenol
Show More Show Less
Hazard Category H227-H302-H310+H330-H314-H317-H335
Hazard Statement GHS H Statement
H311-H330-H314-H318-H227-H302-H317
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P210-P235-P260-P262-P264b-P270-P271-P272-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 2-DIMETHYLAMINOETHYL ACRYLATE
EINECSNumber 219-460-0
RTECSNumber AS8578000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only