missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)-6-propionylnaphthalene 98.0+%, TCI America™

Catalog No. p-7128404
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4678200MG 200 mg
D46781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4678200MG Supplier TCI America Supplier No. D4678200MG

Chemical Identifiers

CAS 70504-01-7
Molecular Formula C15H17NO
Molecular Weight (g/mol) 227.31
MDL Number MFCD00056615
InChI Key MPPQGYCZBNURDG-UHFFFAOYSA-N
Synonym Prodan
PubChem CID 107729
ChEBI CHEBI:51909
IUPAC Name 1-[6-(dimethylamino)naphthalen-2-yl]propan-1-one
SMILES CCC(=O)C1=CC=C2C=C(C=CC2=C1)N(C)C

Specifications

Melting Point 138°C
Color Yellow
Quantity 200 mg
Formula Weight 227.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Dimethylamino)-6-propionylnaphthalene