accessibility menu, dialog, popup

UserName

CAS RN 70504-01-7

IUPAC Name: 1-[6-(dimethylamino)naphthalen-2-yl]propan-1-one
Synonyms: prodan
Molecular Weight (g/mol): 227.31
Molecular Formula: C15H17NO
InChi Key: MPPQGYCZBNURDG-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C2C=C(C=CC2=C1)N(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results