missing translation for 'onlineSavingsMsg'
Learn More

2-(Difluoromethyl)benzimidazole 98.0+%, TCI America™

Catalog No. D47635G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47635G 5 g
D47631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47635G Supplier TCI America Supplier No. D47635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 705-09-9
Molecular Formula C8H6F2N2
Molecular Weight (g/mol) 168.147
MDL Number MFCD00463015
InChI Key PURNIHSRWGYONZ-UHFFFAOYSA-N
PubChem CID 597703
IUPAC Name 2-(difluoromethyl)-1H-benzimidazole
SMILES C1=CC=C2C(=C1)NC(=N2)C(F)F

Specifications

Melting Point 157°C
Quantity 5 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Difluoromethyl)benzimidazole