accessibility menu, dialog, popup

UserName

CAS RN 705-09-9

IUPAC Name: 2-(difluoromethyl)-1H-1,3-benzodiazole
Synonyms: 2-(difluoromethyl)-1H-1,3-benzodiazole
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H6F2N2
InChi Key: PURNIHSRWGYONZ-UHFFFAOYSA-N
SMILES: FC(F)C1=NC2=CC=CC=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)