missing translation for 'onlineSavingsMsg'
Learn More

2-(Difluoromethoxy)benzaldehyde 98.0+%, TCI America™

Catalog No. D48375G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48375G 5 g
D48371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48375G Supplier TCI America Supplier No. D48375G
Only null left

Chemical Identifiers

CAS 71653-64-0
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00042251
InChI Key QPBNHDFPMRENBC-UHFFFAOYSA-N
Synonym 2-difluoromethoxy benzaldehyde, 2-difluoromethoxy-benzaldehyde, o-difluoromethoxybenzaldehyde, 2-difloromethoxybenzaldehyde, pubchem2906, acmc-209ojk, difluoromethoxybenzaldehyde, ksc356m9l, zerenex zx009675, benzaldehyde, difluoromethoxy
PubChem CID 2736984
IUPAC Name 2-(difluoromethoxy)benzaldehyde
SMILES C1=CC=C(C(=C1)C=O)OC(F)F

Specifications

Boiling Point 83°C
Quantity 5 g
Formula Weight 172.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Difluoromethoxy)benzaldehyde
RTECSNumber CU5615600
TSCA No