missing translation for 'onlineSavingsMsg'
Learn More

2-(Diethylamino)ethyl Methacrylate (stabilized with MEHQ) 98.5+%, TCI America™

Catalog No. M0562500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0562500ML 500 mL
M056225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0562500ML Supplier TCI America Supplier No. M0562500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105-16-8
Molecular Formula C10H19NO2
Molecular Weight (g/mol) 185.267
MDL Number MFCD00038314
InChI Key SJIXRGNQPBQWMK-UHFFFAOYSA-N
Synonym Methacrylic Acid 2-(Diethylamino)ethyl Ester
PubChem CID 61012
IUPAC Name 2-(diethylamino)ethyl 2-methylprop-2-enoate
SMILES CCN(CC)CCOC(=O)C(=C)C

Specifications

Color Colorless
Boiling Point 80°C
Quantity 500 mL
Formula Weight 185.27
Percent Purity ≥98.5% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Diethylamino)ethyl Methacrylate (stabilized with MEHQ)
EINECSNumber (2)-1048
RTECSNumber OZ4150000
TSCA Yes