CAS RN 105-16-8
IUPAC Name:
2-(diethylamino)ethyl 2-methylprop-2-enoate
Synonyms:
diethylaminoethyl methacrylate
Molecular Weight (g/mol):
185.27
Molecular Formula:
C10H19NO2
InChi Key:
SJIXRGNQPBQWMK-UHFFFAOYSA-N
SMILES:
CCN(CC)CCOC(=O)C(C)=C
1
–
1
of
1
results
1
2-(Diethylamino)ethyl Methacrylate (stabilized with MEHQ) 98.5+%, TCI America™
CAS: 105-16-8 Molecular Formula: C10H19NO2 Molecular Weight (g/mol): 185.267 MDL Number: MFCD00038314 InChI Key: SJIXRGNQPBQWMK-UHFFFAOYSA-N Synonym: Methacrylic Acid 2-(Diethylamino)ethyl Ester PubChem CID: 61012 IUPAC Name: 2-(diethylamino)ethyl 2-methylprop-2-enoate SMILES: CCN(CC)CCOC(=O)C(=C)C