missing translation for 'onlineSavingsMsg'
Learn More

2-(Diethylamino)ethyl chloride hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog No. p-7022366
Change view
Click to view available options
Quantity:
2500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA151070E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA151070E Supplier Thermo Scientific Chemicals Supplier No. A151070E

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 869-24-9
Molecular Formula C6H15Cl2N
Molecular Weight (g/mol) 172.09
MDL Number MFCD00012519
InChI Key RAGSWDIQBBZLLL-UHFFFAOYSA-N
Synonym 2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride
PubChem CID 13363
SMILES [H+].[Cl-].CCN(CC)CCCl

Specifications

Melting Point 206°C to 210°C
pH 5
Linear Formula (CH3CH2)2NCH2CH2Cl·HCl
Quantity 2500 g
UN Number UN2928
Beilstein 3591991
Sensitivity Moisture sensitive
IUPAC Name 2-chloro-N,N-diethylethanamine;hydrochloride
Formula Weight 172.1
Percent Purity ≥98%
Chemical Name or Material 2-(Diethylamino)ethyl chloride hydrochloride
Show More Show Less
Hazard Category H300+H310+H330-H314-H335
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318-H341
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 212-786-4
RTECSNumber YE1050000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only