accessibility menu, dialog, popup

UserName

HClH3CNH3CCl

CAS RN 869-24-9

IUPAC Name: hydrogen (2-chloroethyl)diethylamine chloride
Synonyms: hydrogen N,N-diethyl-2-chloroethylamine chloride
Molecular Weight (g/mol): 172.09
Molecular Formula: C6H15Cl2N
InChi Key: RAGSWDIQBBZLLL-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCN(CC)CCCl
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)