missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Dicyclohexylphosphino)biphenyl 98.0+%, TCI America™
$72.70 - $260.97
Chemical Identifiers
| CAS | 247940-06-3 |
|---|---|
| Molecular Formula | C24H31P |
| Molecular Weight (g/mol) | 350.486 |
| MDL Number | MFCD01862441 |
| InChI Key | LCSNDSFWVKMJCT-UHFFFAOYSA-N |
| Synonym | 2-dicyclohexylphosphino biphenyl, cyjohnphos, 1,1'-biphenyl-2-yldicyclohexylphosphine, 2-biphenyl dicyclohexylphosphine, 2-biphenylyl dicyclohexylphosphine, biphenyl-2-yldicyclohexylphosphine, dicyclohexyl 2-phenylphenyl phosphane, cyclohexyl johnphos, dicyclohexyl-2-phenylphenyl phosphane, 2-dicyclohexylphosphino-biphenyl |
| PubChem CID | 2734216 |
| IUPAC Name | dicyclohexyl-(2-phenylphenyl)phosphane |
| SMILES | C1CCC(CC1)P(C2CCCCC2)C3=CC=CC=C3C4=CC=CC=C4 |
Chemical Identifiers
| 247940-06-3 | |
| 350.486 | |
| LCSNDSFWVKMJCT-UHFFFAOYSA-N | |
| 2734216 | |
| C1CCC(CC1)P(C2CCCCC2)C3=CC=CC=C3C4=CC=CC=C4 |
| C24H31P | |
| MFCD01862441 | |
| 2-dicyclohexylphosphino biphenyl, cyjohnphos, 1,1'-biphenyl-2-yldicyclohexylphosphine, 2-biphenyl dicyclohexylphosphine, 2-biphenylyl dicyclohexylphosphine, biphenyl-2-yldicyclohexylphosphine, dicyclohexyl 2-phenylphenyl phosphane, cyclohexyl johnphos, dicyclohexyl-2-phenylphenyl phosphane, 2-dicyclohexylphosphino-biphenyl | |
| dicyclohexyl-(2-phenylphenyl)phosphane |
Specifications
| 247940-06-3 | |
| White | |
| MFCD01862441 | |
| 2-dicyclohexylphosphino biphenyl, cyjohnphos, 1,1'-biphenyl-2-yldicyclohexylphosphine, 2-biphenyl dicyclohexylphosphine, 2-biphenylyl dicyclohexylphosphine, biphenyl-2-yldicyclohexylphosphine, dicyclohexyl 2-phenylphenyl phosphane, cyclohexyl johnphos, dicyclohexyl-2-phenylphenyl phosphane, 2-dicyclohexylphosphino-biphenyl | |
| C1CCC(CC1)P(C2CCCCC2)C3=CC=CC=C3C4=CC=CC=C4 | |
| 350.486 | |
| 350.49 | |
| Crystalline Powder |
| 104°C | |
| C24H31P | |
| 1 g | |
| LCSNDSFWVKMJCT-UHFFFAOYSA-N | |
| dicyclohexyl-(2-phenylphenyl)phosphane | |
| 2734216 | |
| ≥98.0% (GC) | |
| 2-(Dicyclohexylphosphino)biphenyl |
Safety and Handling
TSCA : No