accessibility menu, dialog, popup

UserName

P

CAS RN 247940-06-3

IUPAC Name: {[1,1'-biphenyl]-2-yl}dicyclohexylphosphane
Synonyms: [1,1'-biphenyl]-2-yldicyclohexylphosphane
Molecular Weight (g/mol): 350.49
Molecular Formula: C24H31P
InChi Key: LCSNDSFWVKMJCT-UHFFFAOYSA-N
SMILES: C1CCC(CC1)P(C1CCCCC1)C1=CC=CC=C1C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results