missing translation for 'onlineSavingsMsg'
Learn More

2-(Dicyclohexylphosphino)biphenyl 98.0+%, TCI America™

Catalog No. D33885G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33885G 5 g
D33881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33885G Supplier TCI America Supplier No. D33885G
Only null left

Chemical Identifiers

CAS 247940-06-3
Molecular Formula C24H31P
Molecular Weight (g/mol) 350.486
MDL Number MFCD01862441
InChI Key LCSNDSFWVKMJCT-UHFFFAOYSA-N
Synonym 2-dicyclohexylphosphino biphenyl, cyjohnphos, 1,1'-biphenyl-2-yldicyclohexylphosphine, 2-biphenyl dicyclohexylphosphine, 2-biphenylyl dicyclohexylphosphine, biphenyl-2-yldicyclohexylphosphine, dicyclohexyl 2-phenylphenyl phosphane, cyclohexyl johnphos, dicyclohexyl-2-phenylphenyl phosphane, 2-dicyclohexylphosphino-biphenyl
PubChem CID 2734216
IUPAC Name dicyclohexyl-(2-phenylphenyl)phosphane
SMILES C1CCC(CC1)P(C2CCCCC2)C3=CC=CC=C3C4=CC=CC=C4

Specifications

Melting Point 104°C
Color White
Quantity 5 g
Formula Weight 350.49
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Dicyclohexylphosphino)biphenyl