missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl 98.0+%, TCI America™
$186.07 - $546.80
Chemical Identifiers
| CAS | 564483-18-7 |
|---|---|
| Molecular Formula | C33H49P |
| Molecular Weight (g/mol) | 476.729 |
| MDL Number | MFCD04117682 |
| InChI Key | UGOMMVLRQDMAQQ-UHFFFAOYSA-N |
| Synonym | x-phos, 2-dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl, xphos, 2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl, dicyclohexyl 2',4',6'-triisopropyl-1,1'-biphenyl-2-yl phosphine, unii-r271fu23t8, 2-dicyclohexylphosphino-2,4,6-tri-i-propyl-1,1-biphenyl, 2-dicyclohexylphosphino-2',4',6'-tri-isopropyl-1,1'-biphenyl, chemdose tm , x-phos |
| PubChem CID | 11155794 |
| IUPAC Name | dicyclohexyl-[2-[2,4,6-tri(propan-2-yl)phenyl]phenyl]phosphane |
| SMILES | CC(C)C1=CC(=C(C(=C1)C(C)C)C2=CC=CC=C2P(C3CCCCC3)C4CCCCC4)C(C)C |
Chemical Identifiers
| 564483-18-7 | |
| 476.729 | |
| UGOMMVLRQDMAQQ-UHFFFAOYSA-N | |
| 11155794 | |
| CC(C)C1=CC(=C(C(=C1)C(C)C)C2=CC=CC=C2P(C3CCCCC3)C4CCCCC4)C(C)C |
| C33H49P | |
| MFCD04117682 | |
| x-phos, 2-dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl, xphos, 2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl, dicyclohexyl 2',4',6'-triisopropyl-1,1'-biphenyl-2-yl phosphine, unii-r271fu23t8, 2-dicyclohexylphosphino-2,4,6-tri-i-propyl-1,1-biphenyl, 2-dicyclohexylphosphino-2',4',6'-tri-isopropyl-1,1'-biphenyl, chemdose tm , x-phos | |
| dicyclohexyl-[2-[2,4,6-tri(propan-2-yl)phenyl]phenyl]phosphane |
Specifications
| 564483-18-7 | |
| White | |
| MFCD04117682 | |
| x-phos, 2-dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl, xphos, 2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl, dicyclohexyl 2',4',6'-triisopropyl-1,1'-biphenyl-2-yl phosphine, unii-r271fu23t8, 2-dicyclohexylphosphino-2,4,6-tri-i-propyl-1,1-biphenyl, 2-dicyclohexylphosphino-2',4',6'-tri-isopropyl-1,1'-biphenyl, chemdose tm , x-phos | |
| CC(C)C1=CC(=C(C(=C1)C(C)C)C2=CC=CC=C2P(C3CCCCC3)C4CCCCC4)C(C)C | |
| 476.729 | |
| 476.73 | |
| Crystalline Powder |
| 188°C | |
| C33H49P | |
| 1 g | |
| UGOMMVLRQDMAQQ-UHFFFAOYSA-N | |
| dicyclohexyl-[2-[2,4,6-tri(propan-2-yl)phenyl]phenyl]phosphane | |
| 11155794 | |
| ≥98.0% (GC) | |
| 2-Dicyclohexylphosphino-2′,4′,6′-triisopropylbiphenyl |
Safety and Handling
TSCA : No