accessibility menu, dialog, popup

UserName

CAS RN 564483-18-7

IUPAC Name: dicyclohexyl[2',4',6'-tris(propan-2-yl)-[1,1'-biphenyl]-2-yl]phosphane
Synonyms: xphos
Molecular Weight (g/mol): 476.73
Molecular Formula: C33H49P
InChi Key: UGOMMVLRQDMAQQ-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)C1=CC=CC=C1P(C1CCCCC1)C1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results