missing translation for 'onlineSavingsMsg'
Learn More

2-Dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl 98.0+%, TCI America™

Catalog No. D50381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50381G 1 g
D50385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50381G Supplier TCI America Supplier No. D50381G

Chemical Identifiers

CAS 564483-18-7
Molecular Formula C33H49P
Molecular Weight (g/mol) 476.729
MDL Number MFCD04117682
InChI Key UGOMMVLRQDMAQQ-UHFFFAOYSA-N
Synonym x-phos, 2-dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl, xphos, 2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl, dicyclohexyl 2',4',6'-triisopropyl-1,1'-biphenyl-2-yl phosphine, unii-r271fu23t8, 2-dicyclohexylphosphino-2,4,6-tri-i-propyl-1,1-biphenyl, 2-dicyclohexylphosphino-2',4',6'-tri-isopropyl-1,1'-biphenyl, chemdose tm , x-phos
PubChem CID 11155794
IUPAC Name dicyclohexyl-[2-[2,4,6-tri(propan-2-yl)phenyl]phenyl]phosphane
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)C2=CC=CC=C2P(C3CCCCC3)C4CCCCC4)C(C)C

Specifications

Melting Point 188°C
Color White
Quantity 1 g
Formula Weight 476.73
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Dicyclohexylphosphino-2′,4′,6′-triisopropylbiphenyl