missing translation for 'onlineSavingsMsg'
Learn More

2-(Dicyanomethylene)indan-1,3-dione 98.0+%, TCI America™

Catalog No. D20905G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D20905G Supplier TCI America Supplier No. D20905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 90522-66-0
Molecular Formula C12H4N2O2
Molecular Weight (g/mol) 208.176
MDL Number MFCD00051930
InChI Key WQFLMIPTYDGFOB-UHFFFAOYSA-N
PubChem CID 542782
IUPAC Name 2-(1,3-dioxoinden-2-ylidene)propanedinitrile
SMILES C1=CC=C2C(=C1)C(=O)C(=C(C#N)C#N)C2=O

Specifications

Quantity 5 g
UN Number 3439
Formula Weight 208.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Dicyanomethylene)indan-1,3-dione