accessibility menu, dialog, popup

UserName

CAS RN 90522-66-0

IUPAC Name: 2-(1,3-dioxo-2,3-dihydro-1H-inden-2-ylidene)propanedinitrile
Synonyms: 2-(1,3-dioxoinden-2-ylidene)propanedinitrile
Molecular Weight (g/mol): 208.18
Molecular Formula: C12H4N2O2
InChi Key: WQFLMIPTYDGFOB-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C(=O)C1=C(C#N)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results