missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxythymidine-5'-diphosphate trisodium salt, 97%

Catalog No. p-7043005
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64938MB 25 mg
AAJ64938MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64938MB Supplier Thermo Scientific Chemicals Supplier No. J64938MB
Only null left

CAS: 95648-78-5 | C10H13N2Na3O11P2 | 468.13 g/mol

2'-Deoxythymidine-5'-diphosphate trisodium salt is used as a stable nucleotide useful for PCR and RT-PCR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxythymidine-5′-diphosphate trisodium salt is used as a stable nucleotide useful for PCR and RT-PCR.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 95648-78-5
Molecular Formula C10H13N2Na3O11P2
Molecular Weight (g/mol) 468.13
MDL Number MFCD00084748
InChI Key HBYPEEZBYHQLDM-SPSULGLQSA-K
PubChem CID 133109123
IUPAC Name trisodium;[[(2S,3R,5S)-3-hydroxy-5-(5-methyl-2,4-dioxopyrimidin-1-yl)oxolan-2-yl]methoxy-oxidophosphoryl] phosphate
SMILES [Na+].[Na+].[Na+].CC1=CN([C@H]2C[C@H](O)[C@@H](COP([O-])(=O)OP([O-])([O-])=O)O2)C(=O)NC1=O

Specifications

Color White
Quantity 25 mg
Solubility Information Soluble in water.
Formula Weight 468.13
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2'-Deoxythymidine-5'-diphosphate trisodium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only