missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyinosine

Catalog No. AAJ6141109
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6141109 10 g
AAJ6141114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6141109 Supplier Thermo Scientific Chemicals Supplier No. J6141109

CAS: 890-38-0 | C10H12N4O4 | 252.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 890-38-0
Molecular Formula C10H12N4O4
Molecular Weight (g/mol) 252.23
MDL Number MFCD00005762
InChI Key VGONTNSXDCQUGY-YUZWJPFSNA-N
Synonym 2'-deoxyinosine, deoxyinosine, inosine, 2'-deoxy, 2-deoxyinosine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, unii-hn0rq6sbwq, delta-ino, d-ino, hn0rq6sbwq, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine
PubChem CID 65058
ChEBI CHEBI:28997
IUPAC Name 9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1N=CNC2=O

Specifications

Melting Point >250°C
Color White
Quantity 10 g
Beilstein 33517
Formula Weight 252.23
Physical Form Crystalline Powder
Chemical Name or Material 2'-Deoxyinosine
EINECSNumber 212-964-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only