missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 2'-Deoxyinosine, 98+%
CAS: 890-38-0 | C10H12N4O4 | 252.23 g/mol
Supplier: Thermo Scientific Chemicals H5229206
| Quantity | 5 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 890-38-0 | |
| 252.23 | |
| VGONTNSXDCQUGY-YUZWJPFSNA-N | |
| 65058 | |
| 9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one |
| C10H12N4O4 | |
| MFCD00005762 | |
| 2'-deoxyinosine, deoxyinosine, inosine, 2'-deoxy, 2-deoxyinosine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, unii-hn0rq6sbwq, delta-ino, d-ino, hn0rq6sbwq, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine | |
| CHEBI:28997 | |
| OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1N=CNC2=O |
Specifications
| 2'-Deoxyinosine | |
| C10H12N4O4 | |
| MFCD00005762 | |
| VGONTNSXDCQUGY-YUZWJPFSNA-N | |
| 9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one | |
| 65058 | |
| 252.23 |
| 890-38-0 | |
| 5 g | |
| 2'-deoxyinosine, deoxyinosine, inosine, 2'-deoxy, 2-deoxyinosine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, unii-hn0rq6sbwq, delta-ino, d-ino, hn0rq6sbwq, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine | |
| OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1N=CNC2=O | |
| 252.23 | |
| CHEBI:28997 | |
| ≥98% |
Safety and Handling
EINECSNumber : 212-964-1
TSCA : No
Recommended Storage : Ambient temperatures