missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxyguanosine hydrate, 99+%

CAS: 961-07-9 | C10H13N5O4 | 267.25 g/mol

Supplier:  Thermo Scientific Chemicals 226730050

Quantity 5 g

 View more versions of this product

Catalog No. AC226730050


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

961-07-9
267.25
YKBGVTZYEHREMT-DWFKHXNSNA-N
187790
2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
C10H13N5O4
MFCD00150760
2'-deoxyguanosine, deoxyguanosine, guanine deoxyriboside, 2-deoxyguanosine, guanosine, 2'-deoxy, guanine deoxy nucleoside, 2'-deoxy-guanosine, 2;-deoxyguanosine, unii-g9481n71ro, guanine, 9-2-deoxy-beta-d-erythro-pentofuranosyl
CHEBI:17172
NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1

Specifications

2′-Deoxyguanosine hydrate
961-07-9
1.12 to 1.18 (A250/A260) 0.65 to 0.69 (A280/A260)
300°C
(on dry substance) :
Authentic
Glass Bottle
5 g
2'-deoxyguanosine, deoxyguanosine, guanine deoxyriboside, 2-deoxyguanosine, guanosine, 2'-deoxy, guanine deoxy nucleoside, 2'-deoxy-guanosine, 2;-deoxyguanosine, unii-g9481n71ro, guanine, 9-2-deoxy-beta-d-erythro-pentofuranosyl
YKBGVTZYEHREMT-DWFKHXNSNA-N
2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
187790
267.12
Powder
99+%
99.0
100.0
White
300°C
99% min. (HPLC)
C10H13N5O4
MFCD00150760
98% min. (lambda max. 252.5nm,pH 7) molar absorptivity 13700+/-4%,on dry basis
NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1
267.25
CHEBI:17172
99+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 213-505-8

RTECSNumber : MF8760000

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions