missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxyguanosine hydrate, 99+%

Catalog No. p-99054
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC226731000 100 mg
AC226730010 1 g
AC226730050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC226731000 Supplier Thermo Scientific Chemicals Supplier No. 226731000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 961-07-9 | C10H13N5O4 | 267.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 961-07-9
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00150760
InChI Key YKBGVTZYEHREMT-DWFKHXNSNA-N
Synonym 2'-deoxyguanosine, deoxyguanosine, guanine deoxyriboside, 2-deoxyguanosine, guanosine, 2'-deoxy, guanine deoxy nucleoside, 2'-deoxy-guanosine, 2;-deoxyguanosine, unii-g9481n71ro, guanine, 9-2-deoxy-beta-d-erythro-pentofuranosyl
PubChem CID 187790
ChEBI CHEBI:17172
IUPAC Name 2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1

Specifications

CAS Min % 99.0
Absorbance 1.12 to 1.18 (A250/A260) 0.65 to 0.69 (A280/A260)
CAS Max % 100.0
Melting Point 300°C
Color White
Elemental Analysis (on dry substance) :
Flash Point 300°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Spectrophotometry 98% min. (lambda max. 252.5nm,pH 7) molar absorptivity 13700+/-4%,on dry basis
Formula Weight 267.12
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 2′-Deoxyguanosine hydrate, 99+%
Show More Show Less
EINECSNumber 213-505-8
RTECSNumber MF8760000
TSCA TSCA

RUO – Research Use Only