missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxycytidine Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America D004825G

Quantity 25 g

 View more versions of this product

Catalog No. D004825G


Only null left

Description

Description

Specifications

Chemical Identifiers

3992-42-5
263.68
LTKCXZGFJFAPLY-VOBHXVAMNA-N
107488
Cl.NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1
C9H14ClN3O4
MFCD00012840
2'-deoxycytidine hydrochloride, deoxycytidine hydrochloride, cytidine, 2'-deoxy-, monohydrochloride, 2'-deoxycytidine hcl, unii-x8fx60e66d, 2'-deoxycytidine monohydrochloride, cytidine, 2'-deoxy-, hydrochloride 1:1, cytosine deoxyriboside hydrochloride, dc.hcl
4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one hydrochloride

Specifications

2′-Deoxycytidine Hydrochloride
White
25 g
2'-deoxycytidine hydrochloride, deoxycytidine hydrochloride, cytidine, 2'-deoxy-, monohydrochloride, 2'-deoxycytidine hcl, unii-x8fx60e66d, 2'-deoxycytidine monohydrochloride, cytidine, 2'-deoxy-, hydrochloride 1:1, cytosine deoxyriboside hydrochloride, dc.hcl
Cl.NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1
263.68
263.68
Crystalline Powder
3992-42-5
C9H14ClN3O4
MFCD00012840
LTKCXZGFJFAPLY-VOBHXVAMNA-N
4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one hydrochloride
107488
≥98.0% (T)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6834

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions