missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 2'-Deoxycytidine hydrochloride, 99%
CAS: 3992-42-5 | C9H14ClN3O4 | 263.68 g/mol
Supplier: Thermo Scientific Chemicals L1415303
| Quantity | 1 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C9H14ClN3O4 | |
| MFCD00012840 | |
| 2'-deoxycytidine hydrochloride, deoxycytidine hydrochloride, cytidine, 2'-deoxy-, monohydrochloride, 2'-deoxycytidine hcl, unii-x8fx60e66d, 2'-deoxycytidine monohydrochloride, cytidine, 2'-deoxy-, hydrochloride 1:1, cytosine deoxyriboside hydrochloride, dc.hcl | |
| 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride |
Specifications
| 2'-Deoxycytidine hydrochloride | |
| ∽170°C (decomposition) | |
| C9H14ClN3O4 | |
| MFCD00012840 | |
| Hygroscopic | |
| LTKCXZGFJFAPLY-VOBHXVAMNA-N | |
| +57° (c=1 in Water) | |
| 263.68 | |
| 263.68 |
| 3992-42-5 | |
| Odorless | |
| 1 g | |
| 3576124 | |
| 2'-deoxycytidine hydrochloride, deoxycytidine hydrochloride, cytidine, 2'-deoxy-, monohydrochloride, 2'-deoxycytidine hcl, unii-x8fx60e66d, 2'-deoxycytidine monohydrochloride, cytidine, 2'-deoxy-, hydrochloride 1:1, cytosine deoxyriboside hydrochloride, dc.hcl | |
| Cl.NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1 | |
| 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride | |
| 107488 | |
| 99% |
Safety and Handling
EINECSNumber : 223-639-9
TSCA : Yes
Recommended Storage : Ambient temperatures