missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxycytidine hydrochloride, 98%

Catalog No. AC458510010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458510010 1 g
AC458510050 5 g
AC458510250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC458510010 Supplier Thermo Scientific Chemicals Supplier No. 458510010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3992-42-5 | C9H14ClN3O4 | 263.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3992-42-5
Molecular Formula C9H14ClN3O4
Molecular Weight (g/mol) 263.68
MDL Number MFCD00012840
InChI Key LTKCXZGFJFAPLY-VOBHXVAMNA-N
Synonym 2'-deoxycytidine hydrochloride, deoxycytidine hydrochloride, cytidine, 2'-deoxy-, monohydrochloride, 2'-deoxycytidine hcl, unii-x8fx60e66d, 2'-deoxycytidine monohydrochloride, cytidine, 2'-deoxy-, hydrochloride 1:1, cytosine deoxyriboside hydrochloride, dc.hcl
PubChem CID 107488
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride
SMILES Cl.NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 263.68
Percent Purity 98%
Chemical Name or Material 2'-Deoxycytidine hydrochloride
EINECSNumber 223-639-9

RUO – Research Use Only