missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxycytidine 5'-Monophosphate Hydrate 98.0+%, TCI America™

Catalog No. D3673100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3673100MG 100 mg
D36731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3673100MG Supplier TCI America Supplier No. D3673100MG

Chemical Identifiers

CAS 1032-65-1
Molecular Formula C9H14N3O7P
Molecular Weight (g/mol) 307.199
InChI Key NCMVOABPESMRCP-SHYZEUOFSA-N
Synonym dcmp, deoxycytidylic acid, deoxycytidine monophosphate, 2'-deoxycytidine-5'-monophosphate, 2'-deoxycytidine-5'-monophosphoric acid, 5'-cytidylic acid, 2'-deoxy, 2'-deoxycytidine 5'-monophosphate, deoxycytidylate, polydeoxycytidylic acid, deoxycytidine-5'-monophosphoric acid
PubChem CID 13945
ChEBI CHEBI:15918
IUPAC Name [(2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)O)O

Specifications

Melting Point 161°C
Color White-Yellow
Quantity 100 mg
Formula Weight 307.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Deoxycytidine 5′-Monophosphate Hydrate
TSCA No
Recommended Storage Refrigerator