missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxycytidine-5'-monophosphate disodium salt, Thermo Scientific Chemicals

Catalog No. p-7043141
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6572403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6572403 Supplier Thermo Scientific Chemicals Supplier No. J6572403

2?-Deoxycytidine 5-monophosphate (dCMP) is used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form dCDP which upon phosphorylation to dCTP supports DNA and RNA biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxycytidine 5-monophosphate (dCMP) is used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form dCDP which upon phosphorylation to dCTP supports DNA and RNA biosynthesis.

Solubility
Soluble in 200 mg plus 4 mL water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.

Chemical Identifiers

CAS 13085-50-2
Molecular Formula C9H14N3Na2O7P
Molecular Weight (g/mol) 353.178
MDL Number MFCD00069774
InChI Key ACIYKIIBZKDNAD-CDNBRZBRSA-N
Synonym 2'-deoxycytidine-5'-monophosphate disodium salt
PubChem CID 131846647
IUPAC Name [(2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium
SMILES C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)O)O.[Na].[Na]

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in 200 mg plus 4 mL water.
Formula Weight 351.16
Physical Form Powder
Chemical Name or Material 2'-Deoxycytidine-5'-monophosphate disodium salt
EINECSNumber 235-995-2
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only