CAS RN 13085-50-2
IUPAC Name:
disodium (2R,3S,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-2-[(hydrogen phosphonatooxy)methyl]oxolan-3-olate
Synonyms:
disodium (2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-2-[(hydrogen phosphonatooxy)methyl]oxolan-3-olate
Molecular Weight (g/mol):
351.16
Molecular Formula:
C9H12N3Na2O7P
InChi Key:
HKNBOJGORNDJOL-CDNBRZBRSA-M
SMILES:
[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@H]1C[C@H]([O-])[C@@H](COP(O)([O-])=O)O1
1
–
1
of
1
results
1
2'-Deoxycytidine-5'-monophosphate disodium salt, Thermo Scientific Chemicals
CAS: 13085-50-2 Molecular Formula: C9H14N3Na2O7P Molecular Weight (g/mol): 353.178 MDL Number: MFCD00069774 InChI Key: ACIYKIIBZKDNAD-CDNBRZBRSA-N Synonym: 2'-deoxycytidine-5'-monophosphate disodium salt PubChem CID: 131846647 IUPAC Name: [(2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium SMILES: C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)O)O.[Na].[Na]