missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Deoxy-D-ribose, 99%

CAS: 533-67-5 | C5H10O4 | 134.13 g/mol

Supplier:  Thermo Scientific Chemicals A1199006

 View more versions of this product

Catalog No. AAA1199006


Only null left
Explore available promotions

Description

Description

2-Deoxy-D-ribose is used as a precursor to deoxyribonucleic acid. It induces apoptosis by inhibiting the synthesis and increasing the efflux of glutathione. Further, it is used in the synthesis of optically active dipyrrolyl alkanols from pyrroles on the surface of montmorillonite KSF clay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Deoxy-D-ribose is used as a precursor to deoxyribonucleic acid. It induces apoptosis by inhibiting the synthesis and increasing the efflux of glutathione. Further, it is used in the synthesis of optically active dipyrrolyl alkanols from pyrroles on the surface of montmorillonite KSF clay.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

2-Deoxy-D-ribose
76°C to 85°C
MFCD00135904
1721978
14,2908
Soluble in water.
−56° (c=1 in water, 24h)
(3S,4R)-3,4,5-trihydroxypentanal
5460005
134.13
533-67-5
C5H10O4
5 g
Hygroscopic
2-deoxy-d-ribose, thyminose, 3s,4r-3,4,5-trihydroxypentanal, deoxyribose, 2-deoxy-d-erythro-pentose, 2-deoxyribose, d-drib, 2-deoxy-d-arabinose, unii-lsw4h01241, 2-deoxy-d-erythropentose
PDWIQYODPROSQH-GLULIZFNNA-N
OC[C@H]1OC(O)C[C@@H]1O
134.13
CHEBI:28816
99%

Chemical Identifiers

533-67-5
134.13
PDWIQYODPROSQH-GLULIZFNNA-N
5460005
(3S,4R)-3,4,5-trihydroxypentanal
C5H10O4
MFCD00135904
2-deoxy-d-ribose, thyminose, 3s,4r-3,4,5-trihydroxypentanal, deoxyribose, 2-deoxy-d-erythro-pentose, 2-deoxyribose, d-drib, 2-deoxy-d-arabinose, unii-lsw4h01241, 2-deoxy-d-erythropentose
CHEBI:28816
OC[C@H]1OC(O)C[C@@H]1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-573-0

RTECSNumber : SB7230000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only