missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2-Deoxy-D-ribose, 99%

Supplier:  Thermo Scientific™ 153280050

Catalog No. AC153280050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Deoxy-D-ribose
533-67-5
100.0
1.0% max. (40°C, 2 hrs) (vacuum)
98.5% min
C5H10O4
MFCD00135904
-57
Solubility in water: soluble. Other solubilities: soluble in pyridine,slightly soluble in alcohol
C(C=O)C(C(CO)O)O
134.13
CHEBI:28816
99%
99%
98.5
0.5% max.
Authentic
Glass bottle
5g
15, 2909
2-deoxy-d-ribose, thyminose, 3s,4r-3,4,5-trihydroxypentanal, deoxyribose, 2-deoxy-d-erythro-pentose, 2-deoxyribose, d-drib, 2-deoxy-d-arabinose, unii-lsw4h01241, 2-deoxy-d-erythropentose
ASJSAQIRZKANQN-CRCLSJGQSA-N
(3S,4R)-3,4,5-trihydroxypentanal
5460005
134.13
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-573-0

RTECSNumber : SB7230000

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions