missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Deoxy-D-ribose, 99%

Catalog No. AAA1199014
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1199014 25 g
AAA1199003 1 g
AAA1199006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1199014 Supplier Thermo Scientific Chemicals Supplier No. A1199014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 533-67-5 | C5H10O4 | 134.13 g/mol

2-Deoxy-D-ribose is used as a precursor to deoxyribonucleic acid. It induces apoptosis by inhibiting the synthesis and increasing the efflux of glutathione. Further, it is used in the synthesis of optically active dipyrrolyl alkanols from pyrroles on the surface of montmorillonite KSF clay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Deoxy-D-ribose is used as a precursor to deoxyribonucleic acid. It induces apoptosis by inhibiting the synthesis and increasing the efflux of glutathione. Further, it is used in the synthesis of optically active dipyrrolyl alkanols from pyrroles on the surface of montmorillonite KSF clay.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 533-67-5
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.13
MDL Number MFCD00135904
InChI Key PDWIQYODPROSQH-GLULIZFNNA-N
Synonym 2-deoxy-d-ribose, thyminose, 3s,4r-3,4,5-trihydroxypentanal, deoxyribose, 2-deoxy-d-erythro-pentose, 2-deoxyribose, d-drib, 2-deoxy-d-arabinose, unii-lsw4h01241, 2-deoxy-d-erythropentose
PubChem CID 5460005
ChEBI CHEBI:28816
IUPAC Name (3S,4R)-3,4,5-trihydroxypentanal
SMILES OC[C@H]1OC(O)C[C@@H]1O

Specifications

Melting Point 76°C to 85°C
Quantity 25 g
Beilstein 1721978
Sensitivity Hygroscopic
Merck Index 14,2908
Solubility Information Soluble in water.
Optical Rotation −56° (c=1 in water, 24h)
Formula Weight 134.13
Percent Purity 99%
Chemical Name or Material 2-Deoxy-D-ribose
EINECSNumber 208-573-0
RTECSNumber SB7230000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only